Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305558

Sigma-Aldrich

MONOCLONAL ANTI-ERK2 antibody produced in mouse

clone 814CT5.4.2, IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

ERK2, MAPK1, Mitogen-activated protein kinase 1, PRKM1, PRKM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

814CT5.4.2, monoclonal

form

buffered aqueous solution

mol wt

41390 Da

species reactivity

mouse, human

technique(s)

western blot: 1:1000

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK1(5594)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwendolyn I Humphreys et al.
PloS one, 9(11), e111975-e111975 (2014-11-06)
17β-estradiol (E2) plays critical roles in a number of target tissues including the mammary gland, reproductive tract, bone, and brain. Although it is clear that E2 reduces inflammation and ischemia-induced damage in the cerebral cortex, the molecular mechanisms mediating the
Xiaobin Cui et al.
International journal of clinical and experimental pathology, 7(5), 2382-2391 (2014-06-27)
Extracellular signal-regulated kinases (ERKs) are activated by the MAPK pathway. ERKs are downstream effectors of the epidermal growth factor receptor (EGFR), which belongs to the receptor tyrosine kinases family. Studies on the activation of the EGFR-ERK pathway in Kazakh patients
Cynthia L Neben et al.
Human molecular genetics, 23(21), 5659-5671 (2014-06-09)
Fibroblast growth factor receptor 2 (FGFR2) promotes osteoprogenitor proliferation and differentiation during bone development, yet how the receptor elicits these distinct cellular responses remains unclear. Analysis of the FGFR2-skeletal disorder bent bone dysplasia syndrome (BBDS) demonstrates that FGFR2, in addition
Ji-Hee Kim et al.
Oncology reports, 31(6), 2708-2712 (2014-05-03)
Protein tyrosine kinase 7 (PTK7) is a catalytically inactive receptor tyrosine kinase that is also known as colon carcinoma kinase-4 (CCK-4). Recent reports have shown that PTK7 plays an important role in carcinogenesis, and it is known to be upregulated
K A Hlavaty et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(7), 1523-1532 (2014-06-10)
Islet transplantation represents a potential cure for type 1 diabetes, yet the clinical approach of intrahepatic delivery is limited by the microenvironment. Microporous scaffolds enable extrahepatic transplantation, and the microenvironment can be designed to enhance islet engraftment and function. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service