Skip to Content
Merck
All Photos(2)

Key Documents

SAB1300513

Sigma-Aldrich

Anti-UBA2 (C-term E616) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBA2(10054)

General description

Ubiquitin is covalently attached to target proteins by a multienzymatic system consisting of E1 (ubiquitin-activating), E2 (ubiquitin-conjugating), and E3 (ubiquitin-ligating) enzymes. NEDD8, a ubiquitin-like protein, is conjugated to proteins in a manner analogous to ubiquitinylation. beta-amyloid precursor protein-binding protein-1 (APPBP1) can bind to NEDD8 in rabbit reticulocyte lysates. However, since APPBP1 shows similarity to only the N-terminal domain of an E1 enzyme, it must interact with a protein showing similarity to the C-terminal region of E1s. By searching sequence databases, a cDNAs encoding UBA3 was identified as the human homolog of yeast Uba3. The predicted 442-amino acid UBA3 protein shares 43% sequence identity with yeast Uba3. In vitro, UBA3 formed a complex with APPBP1 and a thioester linkage with NEDD8. APPBP1/UBA3 complex may function as an E1-like enzyme for the activation of NEDD8.

Immunogen

UBA2 (NP_005490, 606-641)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human UBA2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service