Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300464

Sigma-Aldrich

Anti-WNK1 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Lysine deficient 1, Anti-PRKWNK1, Anti-Protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WNK1(65125)

General description

The WNK1 gene encodes a cytoplasmic serine-threonine kinase expressed in distal nephron.[supplied by OMIM]
WNK1 (WNK lysine deficient protein kinase 1) gene is mapped to human chromosome 12p13.33. The gene is ubiquitously expressed. The encoded protein contain an active PY motif of short linear proline sequence.

Immunogen

WNK1 (Q9H4A3, 2266-2302)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human WNK1.

Biochem/physiol Actions

WNK is known to influence the activity of chloride transporters such as members of the SLC12 family. WNK1 regulates ion transportation in kidney and maintains chloride concentration in the neurons. It maintains cell volume during the change in the solute concentration. Osmotic stress and chloride depletion within the cell upregulates WNK1 (WNK lysine deficient protein kinase 1) expression. Mutations in WNK1 leads to familial hyperkalemic hypertension, a Mendelian disorder characterized with thiazide-sensitive hypertension, hyperkalemia, and normal glomerular filtration rate. It is associated with hereditary syndromes like renal disease pseudohypoaldosteronism type II (PHAII) and the neurological disease hereditary sensory neuropathy 2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Rooryck et al.
European journal of medical genetics, 52(6), 446-449 (2009-09-08)
We describe a patient presenting with developmental delay, patent foramen ovale, moderate short QT interval, and facial dysmorphism including left microtia, preauricular tag and pit, wide left corner of the mouth, and left hemifacial microsomia, fitting with the oculoauriculovertebral spectrum.
Adriana Mercado et al.
American journal of physiology. Cell physiology, 311(1), C54-C66 (2016-05-14)
The K(+)-Cl(-) cotransporters (KCC1-KCC4) encompass a branch of the SLC12 family of electroneutral cation-coupled chloride cotransporters that translocate ions out of the cell to regulate various factors, including cell volume and intracellular chloride concentration, among others. L-WNK1 is an ubiquitously
Alternatively spliced proline-rich cassettes link WNK1 to aldosterone action.
Roy A
The Journal of Clinical Investigation, 125(9), 3433-3448 (2015)
Ankita Roy et al.
The Journal of clinical investigation, 125(9), 3433-3448 (2015-08-05)
The thiazide-sensitive NaCl cotransporter (NCC) is important for renal salt handling and blood-pressure homeostasis. The canonical NCC-activating pathway consists of With-No-Lysine (WNK) kinases and their downstream effector kinases SPAK and OSR1, which phosphorylate NCC directly. The upstream mechanisms that connect
With no lysine L-WNK1 isoforms are negative regulators of the K+-Cl- cotransporters.
Mercado A
American Journal of Physiology. Cell Physiology, 311(1), C54-C66 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service