Skip to Content
Merck
All Photos(1)

Key Documents

PLA0183

Sigma-Aldrich

Rabbit anti-DAXX Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

BING2, CENP-C binding protein, DAP6, Daxx, EAP1, ETS1-associated protein 1, Fas-binding protein, death-associated protein 6, fas death domain-associated protein, hDaxx

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

western blot: 1:2,000-1:10,000

accession no.

NP_001341.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... DAXX(1616)

Immunogen

The epitope recognized by PLA0183 maps to a region between residue 466 and 516 of human death-associated protein 6 using the numbering given in entry NP_001341.1 (GeneID 1616).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Death-associated protein (DAXX) is a multifunctional protein originally identified as a FAS-interacting protein that modulates FAS-induced cell death. The involvement of DAXX in apoptosis has been difficult to understand, and its exact function in this process has only partially been elucidated. DAXX appears to also function as a transcriptional co-repressor and has been found to associate with the SNF2 chromatin-remodeling ATPase, alpha-thalassemia syndrome protein (ATRX), and modulate transcription.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service