Skip to Content
Merck
All Photos(1)

Key Documents

HPA018914

Sigma-Aldrich

Anti-GGNBP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gametogenetin-binding protein 2, Anti-Laryngeal carcinoma-related protein 1, Anti-Protein ZNF403, Anti-ZNF403 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

VLRAYNILIGELDCSKEKGYCAALYEGLRCCPHERHIHVCCETDFIAHLLGRAEPEFAGGRRERHAKTIDIAQEEVLTCLGIHLYERLHRIWQKLRAE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF403(79893)

General description

The gene gametogenetin-binding gene 2 (GGNBP2/LCRG1) is mapped to human chromosome 17q12-21.1. GGNBP2 transcripts are mainly present in heart, pancreas and skeletal muscle. However, high expression of GGNBP2 is also observed in testis.

Immunogen

Gametogenetin-binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Gametogenetin-binding protein-2 (GGNBP2/LCRG1) acts as a tumor suppressor gene. It is down-regulated in primary laryngeal carcinoma and ovarian cancer. GGNBP2 regulates G2/M cell cycle transition by altering expression of genes, such as up-regulation of p21 and down-regulation of MCM2 (Minichromosome maintenance protein 2 homolog). GGNBP2 is suggested to be involved in spermatogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74460

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuqiang Yin et al.
Oncology reports, 32(1), 362-372 (2014-05-21)
Ribonuclease T2 (RNASET2) and gametogenetin binding protein 2 (GGNBP2) are tumor suppressor genes whose expression is downregulated in ovarian and other types of cancer. However, whether the proteins encoded by these genes are associated with drug resistance has rarely been
T Ohbayashi et al.
FEBS letters, 508(3), 341-344 (2001-12-01)
2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD; dioxin), a member of a class of environmental pollutants represented by polychlorinated dibenzo-p-dioxins and dibenzofurans, is one of the most toxic artificial compounds ever developed. In this study, we identified a novel TCDD target gene, DIF-3 (dioxin inducible
Rui Guan et al.
Molecular and cellular biochemistry, 365(1-2), 211-222 (2012-02-22)
ZNF403, also known as GGNBP2 (gametogenetin binding protein 2), is a highly conserved gene implicated in spermatogenesis. However, the exact biological function of ZNF403 is not clear. In this study, we identified the role of ZNF403 in cell proliferation and
Youjun Li et al.
Cancer letters, 209(1), 75-85 (2004-05-18)
Laryngeal carcinoma (LC) is the most commonly diagnosed malignancy and recently the incidence of this disease has increased. By means of the mRNA differential display method we identified a cDNA, Laryngeal carcinoma related gene 1 (LCRG1) which has significantly reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service