Skip to Content
Merck
All Photos(1)

Key Documents

237035

Sigma-Aldrich

Toluene-d8

"100%", 99.96 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD3
CAS Number:
Molecular Weight:
100.19
Beilstein:
1909300
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.96 atom % D

Assay

≥99.0%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

packaging

pk of 10 × 0.5 mL

technique(s)

GC/MS: suitable
NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.494 (lit.)

bp

110 °C (lit.)

mp

-84 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[2H]

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D

InChI key

YXFVVABEGXRONW-JGUCLWPXSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Visible-Light Photochromism of a Donor-Acceptor Type of Dimethyl 2, 3-bis (arylethynyl) fumarate.
Sakamoto R.
International Journal of Chemical Engineering and Applications, 7(1), 52-52 (2016)
Formation of deuterium atoms in the pyrolysis of toluene-d8 behind shock waves. Kinetics of the reaction C7D8+ H ? C7D7H + D
Rao VS and Skinner GB.
The Journal of Physical Chemistry, 88(94), 4362-4365 (1984)
Fourier transform NMR in liquids at high pressure. III. Spin-lattice relaxation in toluene-d8.
Wilbur DJ and Jonas J.
J. Chem. Phys. , 62(7), 2800-2807 (1975)
Freddy H Havaldar et al.
Journal of labelled compounds & radiopharmaceuticals, 58(4), 163-165 (2015-02-19)
A d4 -labeled isotopomer of deferasirox was synthesized as internal standard for use in a LC/mass spectroscopy (MS)/MS method developed for the simultaneous quantitative determination of deferasirox in human serum. d4 -deferasirox was synthesized from d8 -toluene.
Xiaoyong Lu et al.
Molecules (Basel, Switzerland), 19(9), 14292-14303 (2014-09-13)
Gated molecular encapsulations, with baskets of type 1, are postulated to occur by the mechanism in which solvent molecule penetrates the inner space of 1, through one of its apertures, while the residing guest simultaneously departs the cavity. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service