Skip to Content
Merck
All Photos(1)

Key Documents

213411

Sigma-Aldrich

Sodium triethylborohydride solution

1.0 M in THF

Synonym(s):

Sodium triethylborate, Sodium triethylhydridoborate, Sodium triethylhydroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3NaBH
CAS Number:
Molecular Weight:
121.99
Beilstein:
4151241
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.89 g/mL at 25 °C

SMILES string

[Na+].[H][B-](CC)(CC)CC

InChI

1S/C6H16B.Na/c1-4-7(5-2)6-3;/h7H,4-6H2,1-3H3;/q-1;+1

InChI key

YDTZLEUIYNMRLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of alcohols by hydrogenation of ketones catalyzed by an iron pincer complex
  • Preparation of alkali metal boron monohydride complexes
  • Coordinative substitution reactions
  • Ligand exchange reactions

  • Hydridic reagent

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service