Skip to Content
Merck
All Photos(1)

Key Documents

V800309

Sigma-Aldrich

Potassium carbonate

AR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99%
grade:
AR
form:
powder

grade

AR

product line

Vetec

Assay

≥99%

form

powder

pH

11.0-13 (25 °C, 138 g/L)

mp

891 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Y Foo et al.
Bioresource technology, 104, 679-686 (2011-11-22)
This work explores the feasibility of orange peel, a citrus processing biomass as an alternative precursor for preparation of activated carbon (OPAC) via microwave assisted K(2)CO(3) activation. The operational parameters, chemical impregnation ratio, microwave power and irradiation time on the
Carlos Arroniz et al.
Organic letters, 15(4), 910-913 (2013-02-05)
ortho-Arylation of ortho-substituted benzoic acids is a challenging process due to the tendency of the reaction products toward Pd-catalyzed protodecarboxylation. A simple method for preventing decarboxylation in sterically hindered benzoic acids is reported. The method described represents a reliable and
K Y Foo et al.
Bioresource technology, 102(20), 9794-9799 (2011-08-31)
Sunflower seed oil residue, a by-product of sunflower seed oil refining, was utilized as a feedstock for preparation of activated carbon (SSHAC) via microwave induced K(2)CO(3) chemical activation. SSHAC was characterized by Fourier transform infrared spectroscopy, nitrogen adsorption-desorption and elemental
K Y Foo et al.
Bioresource technology, 111, 425-432 (2012-03-06)
Wood sawdust was converted into a high-quality activated carbon (WSAC) via microwave-induced K(2)CO(3) activation. The operational variables including chemical impregnation ratio, microwave power and irradiation time on the carbon yield and adsorption capability were identified. The surface physical characteristics of
Je-Lueng Shie et al.
Bioresource technology, 102(23), 11011-11017 (2011-10-07)
This study examined the effects of additives of potassium carbonate (K2CO3) and zinc oxide (ZnO) on the pyrolysis of waste sunflower-oil cake using a 60 kW pilot-scale plasma torch reactor. The major gaseous products were CO and H2. The productions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service