Skip to Content
Merck
All Photos(1)

Key Documents

90562

Supelco

2-Hexyl-1-decanol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH[(CH2)5CH3]CH2OH
CAS Number:
Molecular Weight:
242.44
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥96.5% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.449 (lit.)

bp

193-197 °C/33 mmHg (lit.)

mp

−21-−15 °C (lit.)

density

0.836 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC(CO)CCCCCC

InChI

1S/C16H34O/c1-3-5-7-9-10-12-14-16(15-17)13-11-8-6-4-2/h16-17H,3-15H2,1-2H3

InChI key

XULHFMYCBKQGEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

318.2 °F - closed cup

Flash Point(C)

159 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service