Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410027

Sigma-Aldrich

Anti-TRIM14 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA0129

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM14(9830)

General description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies and its function has not been determined. Four alternatively spliced transcript variants for this gene have been described. (provided by RefSeq)

Immunogen

TRIM14 (NP_150089.1, 1 a.a. ~ 442 a.a) full-length human protein.

Sequence
MAGAATGSRTPGRSELVEGCGWRCPEHGDRVAELFCRRCRRCVCALCPVLGAHRGHPVGLALEAAVHVQKLSQECLKQLAIKKQQHIDNITQIEDATEKLKANAESSKTWLKGKFTELRLLLDEEEALAKKFIDKNTQLTLQVYREQADSCREQLDIMNDLSNRVWSISQEPDPVQRLQAYTATEQEMQQQMSLGELCHPVPLSFEPVKSFFKGLVEAVESTLQTPLDIRLKESINCQLSDPSSTKPGTLLKTSPSPERSLLLKYARTPTLDPDTMHARLRLSADRLTVRCGLLGSLGPVPVLRFDALWQVLARDCFATGRHYWEVDVQEAGAGWWVGAAYASLRRRGASAAARLGCNRQSWCLKRYDLEYWAFHDGQRSRLRPRDDLDRLGVFLDYEAGVLAFYDVTGGMSHLHTFRATFQEPLYPALRLWEGAISIPRLP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuan Huang et al.
Journal of molecular cell biology, 12(3), 176-189 (2019-05-10)
Endothelial activation by proinflammatory cytokines is closely associated to the pathogenesis of atherosclerosis and other vascular diseases; however, the molecular mechanisms controlling endothelial activation are not fully understood. Here we identify TRIM14 as a new positive regulator of endothelial activation
Qinghu Ye et al.
Regenerative therapy, 20, 138-146 (2022-05-28)
Coronary artery disease (CAD) is one of the main risks of death, which is mainly caused by coronary arteries arteriosclerosis. Circular RNAs (circRNAs) have shown important regulatory roles in cardiovascular diseases. We amid to explore the role of circ_ROBO2 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service