Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406264

Sigma-Aldrich

Anti-SERPINF2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

A2AP, AAP, ALPHA-2-PI, API, PLI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~54.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINF2(5345)

General description

This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

SERPINF2 (AAH31592.1, 1 a.a. ~ 491 a.a) full-length human protein.

Sequence
MVLLWGLLVLSWSCLQGPCSVFSPVSAMEPLGRQLTSGPNQEQVSPLTLLKLGNQEPGGQTALKSPPGVCSRDPTPEQTHRLARAMMAFTADLFSLVAQTSTCPNLILSPLSVALALSHLALGAQNHTLQRLQQVLHAGSGPCLPHLLSRLCQDLGPGAFRLAARMYLQKGFPIKEDFLEQSEQLFGAKPVSLTGKQEDDLANINQWVKEATEGKIQEFLSGLPEDTVLLLLNAIHFQGFWRNKFDPSLTQRDSFHLDEQFTVPVEMMQARTYPLRWFLLEQPEIQVAHFPFKNNMSFVVLVPTHFEWNVSQVLANLSWDTLHPPLVWERPTKVRLPKLYLKHQMDLVATLSQLGLQELFQAPDLRGISEQSLVVSGVQHQSTLELSEVGVEAAAATSIAMSRMSLSSFSVNRPFLFFIFEDTTGLPLFVGSVRNPNPSAPRELKEQQDSPGNKDFLQSLKGFPRGDKLFGPDLKLVPPMEEDYPQFGSPK

Biochem/physiol Actions

α-2 antiplasmin (AAP) is the primary physiological inhibitor of the serine protease plasmin, which is responsible for the dissolution of fibrin clots. In addition to plasmin, it is also an efficient inhibitor of trypsin and chymotrypsin. α-antiplasmin-deficiency is a rare coagulation disorder which allows unrestrained fibrinolytic activity. Individuals with this condition may receive therapeutic A2AP prior to surgery to prevent postoperative hemorrhaging.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide loss of heterozygosity and copy number alteration in esophageal squamous cell carcinoma using the Affymetrix GeneChip Mapping 10 K array.
Hu N
BMC Genomics, 7, 299-299 (2006)
Open-heart surgery in a patient with heterozygous alpha 2-antiplasmin deficiency. Perioperative strategies in the first reported case.
Shahian DM and Levine JD
Chest, 97(6), 1488-1490 (1990)
The kidney is a major site of alpha(2)-antiplasmin production.
Menoud PA
The Journal of Clinical Investigation, 97(11), 2478-2484 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service