C1624
CLK3, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLJ22858, PHCLK3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
64-86 nmol/min·mg
mol wt
~86 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CLK3(1198)
Biochem/physiol Actions
CLK3, also known as CDC-like kinase 3, encodes a serine/threonine type protein kinase with a non-conserved N-terminal domain. A long and short isoform (phclk3 and pclk3/152) result from alternative splicing and coexist in different tissues. The CLK3 protein has the molecular functions of ATP binding, nucleotide binding, protein serine/threonine kinase activity, protein-tyrosine kinase activity and transferase activity and the CLK3 protein localize in both the cytoplasm and nuclear compartment. CLK3 is thought to regulate the intranuclear distribution of the serine/arginine-rich (SR) family of splicing factors.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Hanes et al.
Journal of molecular biology, 244(5), 665-672 (1994-12-16)
Two new human cDNAs, designated phclk2 and phclk3, which have a high identity to the cDNA of the human protein kinase clk, were characterized. Typical features of hclk2 and hclk3 proteins are non-homologous N-terminal regions and the presence of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service