Skip to Content
Merck
All Photos(1)

Key Documents

24-3910

Sigma-Aldrich

Potassium acetate

JIS special grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOK
CAS Number:
Molecular Weight:
98.14
Beilstein:
3595449
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
grade:
JIS special grade
form:
crystalline

grade

JIS special grade

vapor pressure

<0.0000001 hPa ( 25 °C)

Assay

≥97.0%

form

crystalline

availability

available only in Japan

pH

7.0-9.0 (25 °C, 98.2 g/L)

density

1.57 g/cm3 at 25 °C (lit.)

SMILES string

[K+].CC([O-])=O

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hendrik Marks et al.
Genome biology, 16, 149-149 (2015-08-04)
During early embryonic development, one of the two X chromosomes in mammalian female cells is inactivated to compensate for a potential imbalance in transcript levels with male cells, which contain a single X chromosome. Here, we use mouse female embryonic
Fátima Gebauer et al.
Methods in enzymology, 429, 23-33 (2007-10-05)
Classically, Drosophila cell-free translation systems have been used to study the response of the translational machinery to heat shock treatment. We and others have developed optimized Drosophila embryo and ovary extracts, and their use has expanded to the study of
Yuval Erez et al.
The journal of physical chemistry. A, 116(9), 2039-2048 (2012-02-10)
Steady-state and time-resolved emission techniques were employed to study the effect of acetate, a mild base, on the luminescence of curcumin in methanol and ethanol. We found that the steady-state emission intensity as well as the average fluorescence decay time
Mohammad N Rezaei et al.
International journal of food microbiology, 204, 24-32 (2015-04-02)
Succinic acid produced by yeast during bread dough fermentation can significantly affect the rheological properties of the dough. By introducing mutations in the model S288C yeast strain, we show that the oxidative pathway of the TCA cycle and the glyoxylate
Karin Nowikovsky et al.
The Journal of biological chemistry, 279(29), 30307-30315 (2004-05-13)
The yeast open reading frames YOL027 and YPR125 and their orthologs in various eukaryotes encode proteins with a single predicted trans-membrane domain ranging in molecular mass from 45 to 85 kDa. Hemizygous deletion of their human homolog LETM1 is likely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service