71462
pET-48b(+) DNA - Novagen
Novagen′s pET-48b(+) vector is designed for cloning and high-level expression of proteins fused with the 109aa Trx-Tag thioredoxin protein that is cleavable with the human rhinovirus 3C protease.
Synonym(s):
cloning vector, expression vector, vector
All Photos(1)
About This Item
UNSPSC Code:
41106610
NACRES:
NA.51