Skip to Content
Merck
All Photos(2)

Key Documents

05-785

Sigma-Aldrich

Anti-CD3ε Antibody, clone APA1/1

clone APA1/1, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
APA1/1, monoclonal
application:
FACS
ICC
IP
WB
species reactivity:
mouse, human
technique(s):
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
6

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

APA1/1, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD3E(916)

Specificity

CDε

Immunogen

Purified human CD3 epsilon

Application

Clone APA1/1 has been demonstrated to recognize a site on CD3&epsilon: that is exposed upon engagement of the T-Cell Receptor complex. Exposure of this neoepitope precedes CD3 phosphorylation and recruitment and activation of ZAP-70, which initiates the signaling cascade produced by T-cell activation. As such, clone APA1/1 provides the earliest known marker for T-Cell Receptor engagement, and thus T-cell activation.
Detect CD3ε using this Anti-CD3ε Antibody, clone APA1/1 validated for use in FC, IP, WB & IC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

routinely evaluated by immunoblot on RIPA lysates from Jurkat cells

Target description

~23kDa

Linkage

Replaces: 04-460

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide
Format: Purified
Protein G Purified

Storage and Stability

1 year at 2-8°C from date of shipment.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Alarcón et al.
The EMBO journal, 10(4), 903-912 (1991-04-01)
The T cell receptor for antigen (TCR) consists of two glycoproteins containing variable regions (TCR-alpha/beta or TCR-gamma/delta) which are expressed on the cell surface in association with at least four invariant proteins (CD3-gamma, -delta, -epsilon and -zeta). CD3-gamma and CD3-delta
D A Zapata et al.
The Journal of biological chemistry, 274(49), 35119-35128 (1999-11-27)
Mature CD4(+) and CD8(+) T lymphocytes are believed to build and express essentially identical surface alphabeta T-cell receptor-CD3 (TCR.CD3) complexes. However, TCR.CD3 expression has been shown to be more impaired in CD8(+) cells than in CD4(+) cells when CD3gamma is
Diana Gil et al.
Cell, 109(7), 901-912 (2002-07-12)
How membrane receptors initiate signal transduction upon ligand binding is a matter of intense scrutiny. The T cell receptor complex (TCR-CD3) is composed of TCR alpha/beta ligand binding subunits bound to the CD3 subunits responsible for signal transduction. Although it
A Borroto et al.
The Journal of biological chemistry, 273(21), 12807-12816 (1998-05-28)
Assembly of the six-chain T cell antigen receptor-CD3 complex takes place by pairwise interactions. Thus, CD3-epsilon interacts with either CD3-gamma or CD3-delta, and these dimers then associate with the TCR heterodimer (alpha.beta or gamma.delta) and the CD3-zeta homodimer to constitute

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service