Skip to Content
Merck
All Photos(1)

Key Documents

W527408

Sigma-Aldrich

trans-2,3-Epoxybutane

Synonym(s):

trans-β-Butylene oxide, trans-2,3-Dimethyloxirane, trans-2-Butene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
79772
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
biological source:
synthetic

biological source

synthetic

Quality Level

vapor pressure

3.83 psi ( 20 °C)

refractive index

n20/D 1.3730 (lit.)

bp

54-55 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

C[C@H]1O[C@@H]1C

InChI

1S/C4H8O/c1-3-4(2)5-3/h3-4H,1-2H3/t3-,4-/m1/s1

InChI key

PQXKWPLDPFFDJP-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily C Minerath et al.
Environmental science & technology, 43(5), 1386-1392 (2009-04-09)
Polyols and sulfate esters have recently been identified in the secondary organic aerosol (SOA) formed in the photooxidation of biogenic hydrocarbons both in the laboratory and under ambient atmospheric conditions. In the present study, the potential role of the reactions
C de Meester et al.
Journal of applied toxicology : JAT, 2(6), 284-288 (1982-12-01)
The compound 2,3-epoxybutane (2,3-EB) is a direct mutagen for the base-pair-substitution-sensitive strains TA 1530, TA 1535 and TA 100, in the absence and in the presence of metabolic activation by liver S9-mix. Under the same experimental conditions, the 2-methyl derivative
C de Meester
Toxicology letters, 21(3), 255-262 (1984-06-01)
The mutagenicity of 2,3- epoxybutane (2,3-EB) towards Salmonella FFphimurium TA1530 is increased by rat liver S9 mix. This enhancement can be induced by Aroclor 1254 (ARO) or phenobarbitone (PB) pretreatment. Comparative assays with purified subcellular liver fractions demonstrated that cytosol
P Castelain et al.
Mutagenesis, 8(5), 387-393 (1993-09-01)
Four structurally related aliphatic epoxides (1,2-epoxypropane, 1,2-epoxyisobutane, cis- and trans-2,3-epoxybutane) have been tested in the Salmonella/microsome assay, modified for volatile substances, using the strains TA1535 and TA100. The aim of the study was to evaluate the effect of methylation on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service