Skip to Content
Merck
All Photos(1)

Key Documents

707783

Sigma-Aldrich

Adenosine-15N5 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom % 15N, ≥95% (CP)

Synonym(s):

ATP-15N5 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H1415N5Na2O13P3
Molecular Weight:
556.11
UNSPSC Code:
41106305
eCl@ss:
32160414
NACRES:
NA.12

isotopic purity

≥98 atom % 15N

Quality Level

Assay

≥95% (CP)

form

liquid

contains

5 mM Tris buffer

concentration

100 mM (in 5mM Tris HCl / H2O)
100 mM in H2O

technique(s)

bio NMR: suitable

mass shift

M+5

shipped in

dry ice

storage temp.

−20°C

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Molecular weight and empirical formula are based on the free acid.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service