Merck
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2 application for HPLC
Materiales
Aplicación
Referencia del producto
Descripción
Precios
LC/MS Analysis of Clenbuterol In Plasma on Astec® CHIROBIOTIC® T with Phospholipid Removal using HybridSPE® Phospholipid
application for SPE, application for LC-MS
LC/MS Analysis of Clenbuterol Enantiomers in Plasma on Astec® CHIROBIOTIC® T after SPE using HybridSPE®-Phospholipid
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® V
application for HPLC
Estándar
Referencia del producto
Descripción
Precios
Clenbuterol hydrochloride
≥95%
columna analítica
Referencia del producto
Descripción
Precios
Astec® Chirobiotic® T2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
0.1 wt% ammonium trifluoroacetic acid in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm