Apohemoglobin, carboxymethylated, tryptic digest
Materiales
columna analítica
column
Discovery BIO Wide Pore C5, 15 cm × 4.6 mm I.D., 5 μm particles (568422-U)
mobile phase
[A] 0.1% TFA in water:0.1% TFA in acetonitrile, 95:5; [B] 0.1% TFA in water:0.1% TFA in acetonitrile; 50:50
gradient
0-100% B in 65 min
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
50 μL
sample
carboxymethylated apohemoglobin tryptic digest in 50 mM ammonium bicarbonate