Angiotensins
Materiales
columna analítica
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
(A) 5 mM (NH4)H2PO4/NH4OH, pH 7.0; (B) 5 mM (NH4)H2PO4/NH4OH, pH 7.0: CH3CN (50:50, A:B)
gradient
30-60% B in 15 min
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 220 nm