Key Documents
MTCP70255S
MTC Bio ASPIR-8 Resevoir for 8-Channel Pipettes
new
capacity 25 mL, non-sterile, pack of 200 ea (40 x bags 5 ea)
Sinónimos:
cryo tube box, cryo vial marker, reagent dish, reagent holder
About This Item
Código UNSPSC:
41121522
esterilidad
non-sterile
envase
pack of 200 ea (40 x bags 5 ea)
fabricante / nombre comercial
MTC Bio P7025-5S
capacidad
25 mL
Descripción general
Aspir-8 Reservoir, 8-channel only, 25ml sterile, 5/ sterile bag, 200/cs
Características y beneficios
  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners
Información legal
ASPIR-8 is a trademark of MTC Bio
MTC is a trademark of MTC Bio