Documentos clave
EP951020648
Eppendorf® twin.tec PCR Plates 96
Optimized rim for barcodes, skirt, well volume 150 μL, blue, pkg of 300 ea
About This Item
Código UNSPSC:
41106300
Materiales:
blue
conical bottom
polypropylene wells
pocillo volumen:
150 μL
esterilidad:
non-sterile
Características:
lid: no
skirt
Materiales
blue
conical bottom
polypropylene wells
esterilidad
non-sterile
Características
lid: no
skirt
envase
pkg of 300 ea
pocillo volumen
150 μL
idoneidad
suitable for PCR
Descripción general
Eppendorf® twin.tec PCR Plate 96, skirted, 150 µL, PCR clean, blue, 300 plates
  • Optimized rim for barcodes and automated work processes
  • Within SBS footprint recommendations (127.76 × 85.47 mm ±0.25 mm)
  • Stackable
  • Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
  • Low profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes
  • 150 µL max. well volume (when used with cap strips)
  • Available with barcode (upon request), for skirted and semi-skirted only
Características y beneficios
  • Semi-skirted for label and barcode
  • Fits most thermal cyclers
  • 250 µL max. well volume (when used with cap strips)
  • Improved well-to-well tolerance
  • Available with barcode (upon request), for skirted and semi-skirted only
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)