Documentos clave
EP5427738002
Eppendorf® Adapter
holds 1 x 9-15 mL, bore size (large), pack of 2 ea
About This Item
Código UNSPSC:
41103911
NACRES:
NB.82
Características
holds 1 x 9-15 mL
envase
pack of 2 ea
fabricante / nombre comercial
Eppendorf® 5427738002
técnicas
centrifugation: suitable
calibre tamaño
(large)
compatibilidad
for use with Rotor F-35-6-30 (large rotor bore)
Información legal
Eppendorf is a registered trademark of Eppendorf AG