Documentos clave
EP4981203009
Eppendorf® Multipette plus wall mounting device
pkg of 1 ea
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
envase
pkg of 1 ea
fabricante / nombre comercial
Eppendorf® 4981203009
Descripción general
Self adhesive wall bracket to hold Eppendorf Varipette 4720 or Multipette plus. Not compatible with Eppendorf pipette holdar.
Información legal
Eppendorf is a registered trademark of Eppendorf AG