EP4924000231
Eppendorf® Reference 2 G, BASIC, single-channel, variable
volume 2-20 μL, light gray
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Productos recomendados
Características
single-channel
envase
pack of 1 ea
pkg of 1 ea
fabricante / nombre comercial
Eppendorf® 4924000231
volumen
2-20 μL
color
light gray
Descripción general
The name, Reference, stands for extraordinary precision and accuracy, a long service lifes and an ergonomic design. The new Reference 2 boasts these proven premium characteristics with its state-of-the-art technolgy, making it a reliable partner for you and your work. The time saving single-button operation is unrivalled in the new multichannel versions.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)