Skip to Content
Merck
All Photos(1)

Key Documents

13687-U

Supelco

Amberlite MB20 Ion Exchange Resin

mixed bed, >50 mesh, bucket of 1000 g

Synonym(s):

mix mode ion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin hydrogen and hydroxide form, bucket of 1000 g

form

beads

Quality Level

packaging

bucket of 1000 g

parameter

60 °C max. temp.

moisture

46-51% (cation exchanger)
60-72% (anion exchanger)

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

particle size

560-660 μm (anion form)
710-810 μm (cation form)

operating pH

0-14

capacity

1.0 meq/mL by wetted bed volume(anion)
1.9 meq/mL by wetted bed volume

separation technique

mixed mode

Looking for similar products? Visit Product Comparison Guide

Application

Dowex Marathon MR-3 hydrogen and hydroxide form was used in assessing inorganic nitrogen in litter layer and soil using ion-exchange resin bags on Alpkem autoanalyzer and by ion chromatography.
Mixed bed ion exchange resin specially processed resin for condensate polishing; best for in-process and column demineralization.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional diversity indices reveal the impacts of land use intensification on plant community assembly.
Pakeman, Robin J.
Journal of Ecology, 99, 1143-1151 (2011)
Contrasting effects of substrate and fertilizer nitrogen on the early stages of litter decomposition.
Hobbie, Sarah E.
Ecosystems, 8, 644-656 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service