SRP5202
LATS2 (480-1088), GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLJ13161, KPM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~98 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... LATS2(26524)
General description
LATS2 is a serine/threonine protein kinase belonging to the LATS tumor suppressor family. The kinase activity and two conserved domains within LATS2 are responsible for the suppression of tumorigenicity and inhibition of cell growth. LATS2 negatively regulates the cell cycle by controlling G1/S and/or G2/M transition. LATS2 interacts with the centrosomal proteins AURORA A and Ajuba and is required for accumulation of γ-tubulin and spindle formation at the onset of mitosis. LATS2 also interacts with a negative regulator of p53 and may function in a positive feedback loop with p53 that responds to cytoskeleton damage. LATS2 can also function as a co-repressor of androgen-responsive gene expression.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N Yabuta et al.
Genomics, 63(2), 263-270 (2000-02-16)
We have cloned and characterized LATS2, a novel mammalian homologue of the Drosophila tumor suppressor gene lats/warts. Northern blot analysis showed ubiquitous expression of mouse LATS2 (MmLATS2) mRNA, whereas expression of human LATS2 (HsLATS2) mRNA was enhanced in skeletal muscle
Yunfang Li et al.
Oncogene, 22(28), 4398-4405 (2003-07-11)
Lats2 is a new member of the Lats tumor suppressor family. The human LATS2 gene is located at chromosome 13q11-12, which has been shown to be a hot spot (67%) for LOH in nonsmall cell lung cancer. In order to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service