Skip to Content
Merck
All Photos(1)

Key Documents

P5145

Sigma-Aldrich

Protein A–FITC from Staphylococcus aureus

essentially salt-free, lyophilized powder

Synonym(s):

Fluorescein isothiocyanate Protein A, Protein A–Fluorescein Isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

conjugate

FITC conjugate

Quality Level

form

essentially salt-free, lyophilized powder

extent of labeling

≥50 μg FITC per mg solid

technique(s)

immunohistochemistry (frozen sections): 1:50 using human tonsil

capacity

≥4 mg/mg, solid binding capacity (human IgG)

storage temp.

2-8°C

Specificity

Binds IgG only from most mammals, except rat, goat, sheep.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assay for islet cell antibodies. Protein A--monoclonal antibody method.
Srikanta S et al
Diabetes, 34(3), 300-305 (1985)
P Henning et al.
Human gene therapy, 13(12), 1427-1439 (2002-09-07)
The use of adenovirus (Ad) as an efficient and versatile vector for in vivo tumor therapy requires the modulation of its cellular tropism. We previously developed a method to genetically alter the tropism of Ad5 fibers by replacing the fiber
spa typing method for discriminating among Staphylococcus aureus isolates: implications for use of a single marker to detect genetic micro- and macrovariation.
Koreen L et al
Journal of Clinical Microbiology, 42(2), 792-799 (2004)
A simple method for determining the labeling efficiency of fluorescein isothiocyanate products.
R M McKinney et al.
Analytical biochemistry, 14(3), 421-428 (1966-03-01)
Scott D Kobayashi et al.
mBio, 4(5), e00764-e00713 (2013-10-03)
Staphylococcus aureus is a prominent cause of human infections worldwide and is notorious for its ability to acquire resistance to antibiotics. Methicillin-resistant S. aureus (MRSA), in particular, is endemic in hospitals and is the most frequent cause of community-associated bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service