Skip to Content
Merck
All Photos(2)

Key Documents

AV44398

Sigma-Aldrich

Anti-ZDHHC13 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ10852, Anti-FLJ10941, Anti-HIP14L, Anti-HIP3RP, Anti-MGC64994, Anti-Zinc finger, DHHC-type containing 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

mouse, guinea pig, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ZDHHC13(54503)

General description

Zinc finger, DHHC-type containing 13 (ZDHHC13, HIP14L, HIP3RP) is a Huntingtin-interacting protein that mediates the dual functions of palmitoyl acyltransferase and Mg2+ transport qualifying it as a chanzyme. Protein palmitoylation is important for the regulation of important cellular processes such as protein trafficking, stability, and protein-protein interactions. Improper palmitoylation may underlie diseases such as human alopecia, osteoporosis, and amyloidosis and many other neurodegenerative diseases caused by protein misfolding and amyloidosis.

Specificity

Anti-ZDHHC13 polyclonal antibody reacts with chicken, human, mouse, rat, zebrafish, and bovine zinc finger, DHHC-type containing 13 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZDHHC13

Application

Anti-ZDHHC13 polyclonal antibody is used to tag zinc finger, DHHC-type containing 13 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of zinc finger, DHHC-type containing 13 in processes that involve Mg2+ transport and protein palmitoylation.

Biochem/physiol Actions

ZDHHC13 may be involved in the NF-kappa-B signaling pathway.

Sequence

Synthetic peptide located within the following region: MVILLLQHGADPTLIDGEGFSSIHLAVLFQHMPIIAYLISKGQSVNMTDV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service