Skip to Content
Merck
All Photos(2)

Key Documents

I0639

Supelco

Iodipamide

analytical standard, for drug analysis

Synonym(s):

3,3′(Adipoyldiimino)bis(2,4,6-triiodobenzoic acid), Adipiodone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14I6N2O6
CAS Number:
Molecular Weight:
1139.76
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

OC(=O)c1c(I)cc(I)c(NC(=O)CCCCC(=O)Nc2c(I)cc(I)c(C(O)=O)c2I)c1I

InChI

1S/C20H14I6N2O6/c21-7-5-9(23)17(15(25)13(7)19(31)32)27-11(29)3-1-2-4-12(30)28-18-10(24)6-8(22)14(16(18)26)20(33)34/h5-6H,1-4H2,(H,27,29)(H,28,30)(H,31,32)(H,33,34)

InChI key

FFINMCNLQNTKLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David T Fetzer et al.
Journal of computer assisted tomography, 35(6), 716-722 (2011-11-16)
To determine factors affecting liver and bile duct contrast enhancement during computed tomographic cholangiography (CTC) in living-donor transplant candidates. Forty-four candidates underwent preoperative triphasic CT followed by intravenous infusion of 20 mL of iodipamide for CTC. Body size indices and
Raúl León-Barúa
Revista de gastroenterologia del Peru : organo oficial de la Sociedad de Gastroenterologia del Peru, 32(1), 65-67 (2012-04-06)
In the present article, I remind what I presented, in a symposium performed in our Society on the constipation syndrome, in relation with the definition and the determinant factors of this syndrome, and, in addition, with the methods we have
T Masui et al.
Acta radiologica (Stockholm, Sweden : 1987), 39(5), 557-563 (1998-10-02)
To compare MR and CT cholangiography (MRC and CTC) in evaluating the anatomy of the extrahepatic biliary tract and the pathology related to the gallbladder. Twenty-three patients underwent MRC and CTC with a biliary contrast medium for investigation of biliary
A Persson et al.
The British journal of radiology, 78(936), 1078-1085 (2005-12-15)
The purpose of this study was to evaluate the diagnostic potential of prolonged drip infusion CT cholangiography (DIC-CT) using meglumine iotroxate (Biliscopin) and 3D volume rendering in patients with suspected obstructive biliary disease. From a material of 142 patients who
James S Chen et al.
AJR. American journal of roentgenology, 184(1), 70-74 (2004-12-24)
We sought to investigate the concordance between second-order portal venous and biliary tract anatomies using MDCT angiography and MDCT cholangiography. We retrospectively identified 56 living related potential liver donors who underwent both MDCT angiography and MDCT cholangiography. Two reviewers independently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service