Skip to Content
Merck
All Photos(1)

Key Documents

C20

Flow Cup Viscosity Standard

UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 37.22 mPa.s (20 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

37.22 mPa.s(20 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Flow Cup Viscosity Standards can be found on the Paragon Scientific webpage
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke Tao et al.
Stem cell research & therapy, 7, 20-20 (2016-02-03)
Articular cartilage has a limited potential for self-healing. Transplantation of genetically modified progenitor cells like bone marrow-derived mesenchymal stem cells (MSCs) is an attractive strategy to improve the intrinsic repair capacities of damaged articular cartilage. In this study, we examined
Hye Yeon Choi et al.
Nature communications, 11(1), 3084-3084 (2020-06-20)
Tumor-initiating stem-like cells (TICs) are defective in maintaining asymmetric cell division and responsible for tumor recurrence. Cell-fate-determinant molecule NUMB-interacting protein (TBC1D15) is overexpressed and contributes to p53 degradation in TICs. Here we identify TBC1D15-mediated oncogenic mechanisms and tested the tumorigenic
James C Fleet et al.
Cancer prevention research (Philadelphia, Pa.), 12(6), 343-356 (2019-04-28)
We tested whether lifelong modification of vitamin D signaling can alter the progression of early prostate carcinogenesis in studies using mice that develop high-grade prostatic intraepithelial neoplasia that is similar to humans. Two tissue-limited models showed that prostate vitamin D
Nattapong Puthdee et al.
Biological & pharmaceutical bulletin, 40(6), 751-757 (2017-06-02)
Berberine is a natural compound found in several herbs. Anticancer activity of berberine was reported in several cancers, however, little is known regarding the effects of berberine against cholangiocarcinoma (CCA). In this study, the growth inhibitory effects of berberine on
Rosaria Ottanà et al.
European journal of medicinal chemistry, 127, 840-858 (2016-11-16)
New 4-{[5-arylidene-2-(4-fluorophenylimino)-4-oxothiazolidin-3-yl]methyl}benzoic acids (5) and 2-thioxo-4-thiazolidinone analogues (6) were synthesised as a part of a continuing search for new inhibitors of protein tyrosine phosphatase 1B (PTP1B), an enzyme which is implicated in metabolic disorders and inflammatory signaling. Most of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service