Skip to Content
Merck
All Photos(1)

Key Documents

SK1M014W01

Millipore

Pellicon® 2 Cassette with Biomax 5 kDa Membrane, C screen, 0.5 m²

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031623

material

polyethersulfone
polypropylene
polypropylene screen
urethane foam-filled high-density polyethylene

Quality Level

description

Filter code Biomax 5
Type C screen (coarse screen)
prefiltration requirements: 100 µm

quality

400

parameter

0-50 °C max. temp.
0.3 bar max. transmembrane pressure (5 psig) at 30 °C (Reverse)
4-7 L/min-cm2 recirculation rate
6.8 bar max. transmembrane pressure (100 psig) at 30 °C (Forward)

technique(s)

analytical sample preparation: suitable
protein purification: suitable
tangential flow filtration (TFF): suitable
ultrafiltration: suitable

L

20.6 cm (8.1 in.)

membrane nominal area

0.1 m2

width

17.8 cm , 7.0 in.

matrix

Biomax®

pore size

5 kDa filter

pH range

1-14

Analysis Note

The extractables level was equal to or less than 1500 mg/m² after a water flush.

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service