Skip to Content
Merck
All Photos(1)

Key Documents

700191P

Avanti

Oxidized 18:2 cholesterol(d7)

Avanti Research - A Croda Brand (700191P), powder

Synonym(s):

Oxidized cholesteryl(d7) linoleate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 500 μg (700191P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand (700191P)

shipped in

dry ice

storage temp.

−20°C

General description

Cholesteryl linoleate (LA-CE) is a major polyunsaturated fatty acid- containing cholesterol ester. It is found in the core of low density lipoprotein (LDL).

Biochem/physiol Actions

Cholesteryl linoleate (LA-CE) exhibits bactericidal property.

Packaging

5 mL Amber Glass Screw Cap Vial (700191P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipids including cholesteryl linoleate and cholesteryl arachidonate contribute to the inherent antibacterial activity of human nasal fluid
Do Thai Q, et al.
Journal of Immunology, 181(6), 4177-4187 (2008)
Endogenous cholesterol ester hydroperoxides modulate cholesterol levels and inhibit cholesterol uptake in hepatocytes and macrophages
Guo S et al.
Redox Biology, 21, 101069-101069 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service