Skip to Content
Merck
All Photos(1)

Key Documents

GF64169051

T2 - thermocouple alloy, Ni95(Al+Mn+Si)5

insulated wire, 10m length, 0.125mm conductor diameter, 0.004mm insulation thickness, polyester insulation

Synonym(s):

NI105840

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

description

polyester insulation
thermocouple wire

form

wire (insulated)

manufacturer/tradename

Goodfellow 641-690-51

insulation thickness

0.004 mm

wire L × diam.

10 m × 0.125 mm , conductor diameter

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Application

may be used as one component of a thermocouple

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service