GC Analysis of Native Spearmint Essential Oil on SUPELCOWAX 10
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
75 °C (4 min), then 4 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Native Spearmint Oil, (A.M. Todd Sample), neat
Description
Legal Information
null