GC Analysis of JET-A on Equity-1, Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
carrier gas
hydrogen, 45 cm/sec constant
column
Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
detector
FID, 350 °C
inj. temp.
250 °C
injection
0.1 μL, 300:1 split
liner
2 mm I.D., straight
oven
80 °C, 50 °C/min to 325 °C
sample
JET-A, neat
Description
Legal Information
null