GC Analysis of Permanent Gases on Carboxen®-1010 PLOT
![GC Analysis of Permanent Gases on Carboxen®-1010 PLOT suitable for GC](/deepweb/assets/sigmaaldrich/product/images/327/209/8494052f-4bf1-43b9-ae55-a8903f426ced/800/8494052f-4bf1-43b9-ae55-a8903f426ced.jpg)
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
Carbon Monoxide (7%), Carbon Dioxide (15%), Oxygen (4%), and Methane (4.5%).
In in nitrogen, cylinder of 14 L, analytical standardCONDITIONS
column
Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven
35 °C (7.0 min), 24 °C/min to 225 °C
inj. temp.
200 °C
detector
TCD, 230 °C
carrier gas
helium, 3.0 mL/min
injection
10 μL, direct using a sample valve, 150 °C
sample
Oxygen, carbon monoxide, methane, and carbon dioxide, each at 1.0% in nitrogen
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany