Merck
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity-1 after Purge & Trap using "M" Trap
US EPA Method 8015 (modified): GC Analysis of Gasoline Range Organics (GRO) on Equity-1 after Purge & Trap using "M" Trap suitable for GC
Materials
standard
Product No.
Description
Pricing
EPA 8270 Semivolatile Internal Standard Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride, ampule of 2 × 1 mL
CONDITIONS
sample/matrix
Each analyte at 50 ppb in 5 mL of water
purge trap
MOD BTEXTRAP M, Tekmar® 3000
purge
40 mL/min at 25 °C for 11 min
dry purge
2 min
desorption preheat
220 °C
bake
260 °C for 10 min
transfer line/valve temp.
110 °C
column
Equity-1, 30 m x 0.32 mm I.D., 1.0 μm (28057-U)
oven
40 °C (2 min), 5 °C/min to 75 °C, 10 °C/min to 200 °C
inj. temp.
200 °C
detector
FID, 250 °C
carrier gas
helium, 1.4 mL/min constant
injection
20:1 split
liner
0.75 mm I.D. SPME
Description
Legal Information
null
Tekmar is a registered trademark of Tekmar Co.