Skip to Content
Merck
All Photos(1)

Key Documents

818827

TOYOPEARL® AF-Carboxy-650M Bulk Media

phase carboxylic acid, bottle of 1 L, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Carboxy-650M Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
particle size:
65 μm
matrix active group:
carboxylic acid phase
pore size:
1000 Å
matrix:
hydroxylated methacrylic polymer particle platform

description

Adsorption Capaacity (40 ± 10 mg/mL (lysozyme))
Shipping solvent (20% (v/v) ethanol)
phase filter (Carboxy)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 18827

extent of labeling

80-120 μeq/mL ligand density

parameter

pressure (max. 3 bar)

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

carboxylic acid phase

particle size

65 μm

pore size

1000 Å

pH

2-13

capacity

40 g/L±10 adsorption capacity (lysozyme)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Carboxy-650 media is a reactive resin which couples to the amino groups of proteins or smaller molecular weight ligands. The carbodiimide mediated coupling reaction produces an amide bond between the support and the ligand.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service