Z327794
BRAND® graduated pipette, PP, high clarity
capacity 0 (5 mL) , 0.05 mL accuracy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polypropylene
description
blue ring mark
feature
graduations
subdivision 0.1 mL
packaging
pack of 12
manufacturer/tradename
BRAND 27612
capacity
0 (5 mL) , 0.05 mL accuracy
suction tube O.D.
≤8 mm
Looking for similar products? Visit Product Comparison Guide
General description
Resistant to breakage. Ideal for applications where glass pipettes cannot be used, e.g. in fluoro and silicate chemistry. Exposure to temperatures up to 60°C will not cause permanent exceeding of error limits. Cleaning temperatures below 60°C is recommended to preserve marks and inscriptions
Other Notes
BR27612 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service