Z325481
BRAND® SILBERBRAND ETERNA graduated pipette, calibrated to deliver (TD, EX)
capacity 2 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
AR-Glass
feature
class B
graduation
subdivision 0.02 mL
supplementary to DIN range: no
packaging
pack of 12
manufacturer/tradename
BRAND 27073
capacity
2 mL
Looking for similar products? Visit Product Comparison Guide
General description
AR-glass®. Graduated pipettes according to DIN EN ISO 835. Type 3 (zero point at the top), total delivery. Graduations and inscriptions in ETERNA amber stain, particularly resistant to aggressive cleaning methods.
Other Notes
BR27073 (India)
Legal Information
Araldite is a registered trademark of Huntsman Advanced Materials Inc.
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service