Skip to Content
Merck
All Photos(3)

Key Documents

WH0002246M1

Sigma-Aldrich

Monoclonal Anti-FGF1 antibody produced in mouse

clone 3F5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AFGF, Anti-ECGF, Anti-ECGFA, Anti-ECGFB, Anti-ECGFbeta, Anti-FGFA, Anti-FGFalpha, Anti-GLIO703, Anti-HBGF1, Anti-fibroblast growth factor 1 (acidic)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF1(2246)

General description

FGF1 (fibroblast growth factor 1) gene encodes a member of the fibroblast growth factor (FGF) family. It encodes a pro-angiogenic protein that is ubiquitously expressed. In humans, FGF1 is alternatively spliced into four forms: FGF1A (in kidney), FGF1B (in brain), FGF1-C and -D (in vascular smooth muscle cells and fibroblasts). This gene is located on human chromosome 5q31.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis. Multiple alternatively spliced variants encoding different isoforms have been described. (provided by RefSeq)

Immunogen

FGF1 (AAH32697, 46 a.a. ~ 155 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VDGTRDRSDQHIQLQLSAESVGEVYIKSTETGQYLAMDTDGLLYGSQTPNEECLFLERLEENHYNTYISKKHAEKNWFVGLKKNGSCKRGPRTHYGQKAILFLPLPVSSD

Biochem/physiol Actions

FGF1 (fibroblast growth factor 1) is mainly involved in cell growth, proliferation and neurogenesis. This gene also participates in wound healing, post-ischemic heart repair and making of collaterals after hindlimb ischemia. The protein functions as an angiogenic factor that participates in tissue repair, carcinogenesis, and maintenance of vasculature stability.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transgenic expression of nonclassically secreted FGF suppresses kidney repair
Kirov A, et al.
PLoS ONE (2012)
Upregulation of fibroblast growth factor 1 in the synovial membranes of patients with late stage osteoarthritis
Li R, et al.
Genetics and molecular research : GMR (2015)
Regulation of FGF1 gene promoter through transcription factor RFX1
Hsu YC, et al.
The Journal of Biological Chemistry, 285(18), 13885-13895 (2010)
Fibroblast growth factors
Ornitz DM and Itoh N
Genome Biology (2001)
Folding of Fibroblast Growth Factor 1 Is Critical for Its Nonclassical Release
Prudovsky I, et al.
Biochemistry, 55(7), 1159-1167 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service