Skip to Content
Merck
All Photos(1)

Key Documents

SRP0501

Sigma-Aldrich

C-MER, GST-Tag

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

MERTK, receptor tyrosine kinase MerTK, tyrosine-protein kinase Mer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

60 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MERTK(10461)

General description

Human c-Mer, also known as tyrosine-protein kinase Mer, MERTK, and receptor tyrosine kinase MerTK GenBank Accession NM_006343, a.a. 578-872 with C-terminal GST-tag. Expressed in a baculovirus-infected Sf9 cell expression system. MW = 60 kDa.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service