Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411924

Sigma-Aldrich

ANTI-MS4A12 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MS4A12, Ms4a10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 28.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MS4A12(54860)

General description

The protein encoded by this gene is a cell surface protein found primarily in the apical membrane of colonocytes. Silencing of this gene in colon cancer cells inhibits the proliferation, cell motility, and chemotactic invasion of cells. This gene is part of a cluster of similar genes found on chromosome 11. Two transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

MS4A12 (AAH29793.1, 1 a.a. ~ 267 a.a) full-length human protein.

Sequence
MMSSKPTSHAEVNETIPNPYPPSSFMAPGFQQPLGSINLENQAQGAQRAQPYGITSPGIFASSQPGQGNIQMINPSVGTAVMNFKEEAKALGVIQIMVGLMHIGFGIVLCLISFSFREVLGFASTAVIGGYPFWGGLSFIISGSLSVSASKELSRCLVKGSLGMNIVSSILAFIGVILLLVDMCINGVAGQDYWAVLSGKGISATLMIFSLLEFFVACATAHFANQANTTTNMSVLVIPNMYESNPVTPASSSAPPRCNNYSANAPK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service