Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR384M2A

PCR microplate for ABI 3730, 3100 and MJ Thermocyclers

size 384 wells, pkg of 5x10plates/cs, assorted colors

Synonym(s):

384 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
384 wells

material

conical bottom wells

manufacturer/tradename

Axygen Inc PCR-384M2-A
Corning

packaging

pkg of 5x10plates/cs

size

384 wells

color

assorted colors

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service