MABF2745
Anti-CD200R Antibody, clone OX-108
Synonym(s):
CD200 cell surface glycoprotein receptor, CD200R, CRTR2, Cell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, MOX2R, OX2R, UNQ2522/PRO6015
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
OX-108, monoclonal
mol wt
calculated mol wt 36.60 kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100
technique(s)
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
inhibition assay: suitable
surface plasmon resonance (SPR): suitable
isotype
IgG1κ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
human ... CD200R1(131450)
Specificity
Immunogen
Application
Evaluated by Flow Cytometry with human peripheral blood mononuclear cells (PBMCs).
Flow Cytometry Analysis: 1.0 µg from a representative lot detected CD200R in one million human peripheral blood mononuclear cells (PBMCs).
Tested Applications
Surface Plasmon Resonance: A representative lot detected CD200R in surface plasmon resonance application (Hatherley, D., et al. (2004). Eur J Immunol. 34(6):1688-94).
Immunohistochemistry Application: A representative lot detected CD200R in immunohistochemistry application (Koning, N., et al. (2009). J Neuropathol Exp Neurol. 68(2):159-67).
Inhibition: A representative lot blocked ligand-receptor interactions in protein microarray application (Letarte, M., et al. (2005). BMC Biochem.1:6:2).
Flow Cytometry Analysis: A representative lot detected CD200R in flow cytometry application (Wright, G.J., et al. (2003). J Immunol. 171(6):3034-46; Foster-Cuevas, M., et al. (2004). J Virol. 78(14):7667-76; Koning, N., et al. (2009). J Neuropathol Exp Neurol. 68(2):159-67).
Immunofluorescence Analysis: A representative lot detected CD200R in immunofluorescence application (Letarte, M., et al. (2005). BMC Biochem.1:6:2).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service