Skip to Content
Merck
All Photos(1)

Key Documents

MABE1066

Sigma-Aldrich

Anti-SETDB1 Antibody, clone F0838

clone F0838, from mouse

Synonym(s):

Histone-lysine N-methyltransferase SETDB1, EC: 2.1.1.43, ERG-associated protein with SET domain, ESET, SET domain bifurcated 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

F0838, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Setdb1(84505)

General description

Histone-lysine N-methyltransferase SETDB1 (UniProt: O88974; also known as EC: 2.1.1.43, ERG-associated protein with SET domain, ESET, SET domain bifurcated 1) is encoded by the Setdb1 (also known as Eset, Kiaa0067) gene in human. SETDB1 is ubiquitously expressed, but a strong expression is observed in liver and testis. It is a histone methyltransferase that specifically trimethylates Histone 3 Lysine 9 (H3K9). H3K9 trimethylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 (CBX1, CBX3 and/or CBX5) proteins to methylated histones. It mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. SETDB1 is reported to inhibit adipogenesis. It has also been shown that H3K4/H3K9me3 bivalent chromatin signature keeps expression of Cebpa and Pparg genes very low. Depletion of SETDB1 can induce adipogenesis in mesenchymal stem cells and in preadipocytes. SETDB1 contains two Tudor domains (aa 257-320 and 347-403) and one MBD domain (aa 611-682). It also has a SET domain (aa 820-1282), a pre-SET (aa 744-817), and a post-SET domain (aa 1291-1307). The pre-SET, SET and post-SET domains are required for its methyltransferase activity. In the pre-SET domain, Cys residues bind 3 zinc ions that are arranged in a triangular cluster; some of these Cys residues contribute to the binding of two zinc ions within the cluster. (Ref.: Matsumura, Y., et al., (2015), Mol. Cell 60(4); 584- 596).

Specificity

Clone F0838 recognizes SETDB1 in murine embryonic stem cells. It targets an epitope with in 213 amino acids from the N-terminal half of isoform 1 and 7.

Immunogen

His-tagged recombinant fragment corresponding to 213 amino acids from the N-terminal half of murine Histone-lysine N-methyltransferase SETDB1.

Application

Anti-SETDB1, clone F0838, Cat. No. MABE1066, is a highly specific mouse monoclonal antibody that detects SETDB1 and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected SETDB1 in Western Blotting applications (Matsumura, Y, et. al. (2015). Mol Cell. 60(4):584-96).

Quality

Evaluated by Western Blotting in 129/S6/SvEv mouse embryonic stem cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected SETDB1 in 129/S6/SvEv mouse embryonic stem cell lysate.

Target description

~160 and 180 kDa observed; 144.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS with 0.01% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service