MAB331
Anti-SNAP-25 Antibody, clone SP14
ascites fluid, clone SP14, Chemicon®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
mouse
antibody form
ascites fluid
clone
SP14, monoclonal
species reactivity
pig, human, hamster, rat
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... SNAP25(6616)
Specificity
SNAP-25 (26-27 kDa). MAB331 is reactive with fusion protein from COS cells, but not with fusion protein from bacterial systems.
Immunogen
Crude synaptic immunoprecipitate (Human)
Application
Detect SNAP-25 using this Anti-SNAP-25 Antibody, clone SP14 validated for use in ELISA, WB, IH(P).
Immunohistochemistry at 1:500-1:5,000
Formalin fixed (<4 weeks), paraffin embedded
* It is suggested that you use an IgG specific second antibody to minimize possible non-specific staining.
Frozen
Western blotting at 1:100-1:1,000
ELISA at 1:500
It is reported that clone SP14 is capable of detecting SDS-resistant SNARE complexes {J. Cell Sci 2004 117(6):955-966}.
Optimal working dilutions must be determined by the end user.
Formalin fixed (<4 weeks), paraffin embedded
* It is suggested that you use an IgG specific second antibody to minimize possible non-specific staining.
Frozen
Western blotting at 1:100-1:1,000
ELISA at 1:500
It is reported that clone SP14 is capable of detecting SDS-resistant SNARE complexes {J. Cell Sci 2004 117(6):955-966}.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Synapse & Synaptic Biology
Physical form
Liquid. Contains no preservative.
Storage and Stability
Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bronchoepithelial expression of CXCR1 and CXCR2 does not facilitate transepithelial migration of neutrophils.
Respiration; International Review of Thoracic Diseases null
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(17), 4618-4634 (2017-04-02)
G-protein βγ subunits (Gβγ) interact with presynaptic proteins and regulate neurotransmitter release downstream of Ca2+ influx. To accomplish their roles in sensory signaling, photoreceptor synapses use specialized presynaptic proteins that support neurotransmission at active zone structures known as ribbons. While
High sensitivity of mouse neuronal cells to tetanus toxin requires a GPI-anchored protein.
Biochemical and biophysical research communications null
The gene for soluble N-ethylmaleimide sensitive factor attachment protein alpha is mutated in hydrocephaly with hop gait (hyh) mice.
Proceedings of the National Academy of Sciences of the USA null
Most peptide-containing sensory neurons lack proteins for exocytotic release and vesicular transport of glutamate.
The Journal of Comparative Neurology null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service