1.71597
Parteck® SI 400 LEX (Sorbito 2)
Pharma Manufacturing
Synonym(s):
Parteck® Sorbitol
About This Item
Recommended Products
vapor pressure
<0.1 hPa ( 25 °C)
Quality Level
pH
5.0-7 (25 °C, 182 g/L in water)
mp
94-96 °C
solubility
2350 g/L
density
450 kg/m3
SMILES string
OC(C(O)C(O)CO)C(O)CO
InChI
1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2
InChI key
FBPFZTCFMRRESA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Application
also commonly purchased with this product
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service