Skip to Content
Merck
All Photos(1)

Key Documents

GF13169594

Manganin® - resistance alloy, Cu86Mn12Ni2

foil, 100 x 100mm, 0.02mm thickness, as rolled, light tight (lt)

Synonym(s):

CU060220

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

description

light tight (lt)

Assay

99.90%

form

foil (as rolled)

manufacturer/tradename

Goodfellow 131-695-94

size × thickness

100 mm × 100 mm × 0.02 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Manganin is a registered trademark of Isabellenhütte Heusler GmbH & Co. KG.
Manganin is a registered trademark of Isabellenhütte Heusler GmbH & Co. KG.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service